Learn More
Thermo Scientific™ Orion Star™ A121 Portable pH Meter
Take simple pH and temperature measurements with our portable pH meter with easy operation for basic field testing.
$1710.56 - $1833.20
Specifications
Accuracy (mV) | ±0.2 or ±0.05%, whichever is greater |
---|---|
Accuracy (pH) | ±0.01 |
Accuracy (Temperature) | ±0.1°C |
Data Management Details | Internal Data Point Memory |
Outputs | None |
Catalog Number | Mfr. No. | Description | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Includes | Price | Quantity | ||||
13-645-504
|
Thermo Scientific™
STARA1210 |
Orion Star A121 portable pH meter | Star A121 pH meter, 4 AA batteries |
Each for $1,710.56
|
|
||||
13-645-506
|
Thermo Scientific™
STARA1215 |
Orion Star A121 portable pH meter standard kit with general purpose pH/ATC electrode | Star A121 pH meter, 9107BNMD gel-filled epoxy-body pH/ATC electrode, 916099 pH buffer kit, protective meter armor, hard-sided carrying case, 4 AA batteries |
Each for $1,833.20
|
|
||||
Description
Take simple, routine pH, mV and temperature measurements with the Thermo Scientific™ Orion Star™ A121 Portable pH Meter. This meter is an easy-to-use and budget-friendly instrument for basic pH analysis in the field. Perform up to a 3 point pH calibration with easy recall of calibration points and slope. Quickly navigate setup menus and ensure consistent calibrations using the simple keypad layout and on-screen text prompts. This waterproof meter has an IP67-rated housing for rugged protection in outdoor environments.
- Measure pH, mV or relative mV with temperature
- Perform up to a 3 point pH calibration with automatic buffer recognition or manual buffer entry
- Simplified, easy-to-read display shows main measurement with temperature, stability indicator and electrode condition icon
- User-friendly operation with basic calibration text prompts
- Easily recall calibration data for procedure checks
- Non-volatile memory preserves 50 point internal data log and meter settings, even with loss of power
- Flexible power options with AA batteries or optional universal power adapter
- IP67-rated housing is waterproof for rugged field operation
- Meter kits include hard-sided field case and protective meter armor with electrode holders, adjustable wrist/belt strap and built-in meter stand
- Meters offer CE, TUV 3-in-1 and FCC Class A certifications
- 3-year replacement warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
±0.2 or ±0.05%, whichever is greater | |
±0.1°C | |
None | |
pH, relative mV (RmV) and temperature | |
0.01, 0.1 | |
0.1mV | |
-5°C to +105°C | |
0.1°C, 0.1°F | |
1 point temperature offset calibration | |
Non-volatile memory preserves log and settings | |
Automatic with Auto-Read Measure Mode, Manual with Continuous Measure Node | |
CE, TUV 3-1, FCC Class A | |
Universal 50-60 Hz, 100-240 VAC power adapter (sold separately, Cat. No. 1010003) | |
0.5 kg | |
3 Year Meter Warranty | |
Orion Star A121 Portable pH/mV/RmV/Temperature Meter | |
English | |
5 to 85%, non-condensing | |
Smart Stability and Smart Averaging | |
5°C to 45°C (ambient) | |
4 x AA |
±0.01 | |
Internal Data Point Memory | |
pH, mV, Relative mV with Temperature | |
-2 to 16 | |
±1600.0 | |
1 to 3 | |
22.0°F to 221.0°F | |
Manual or automatic with ATC temperature probe | |
LCD | |
50 | |
BNC (pH or ORP electrode), 8 pin MiniDIN (ATC temperature probe) | |
IP-67 | |
24 cm x 10 cm x 6.5 cm (9.5 in. x 3.9 in. x 2.6 in.) | |
2 lb. | |
Simple with highlighted measure key, scroll and shortcut keys | |
4 AA batteries (included) or universal AC adapter (sold separately) | |
±1999.9mV | |
7.00pH | |
41°F to 113°F (ambient) | |
1 | |
2000 hr. |