missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Orion™ Storage Sleeve and Base

Protect electrodes from breakage when not in use with the Thermo Scientific™ Orion™ Electrode Storage Sleeve and Base.

Supplier:  Thermo Scientific™ 810017

Catalog No. 13-642-145


Explore available promotions

Description

Description

Protect electrodes from breakage when not in use with the Thermo Scientific™ Orion™ Electrode Storage Sleeve and Base.

The Thermo Scientific Orion Storage Sleeve and Base is for use with pH electrodes that are 120mm or less in length that have a diameter of 12mm. The sleeve protects your electrode from breakage when not in use and will not tip, due to the weighted removable base. As an additional convenience, the pH bulb stays conditioned while in storage so that it is ready to use when removed from the sleeve.

Specifications

Specifications

Orion Storage Sleeve and Base
pH Electrode
Storage Sleeve, Base
Videos
SDS
Documents

Documents

Promotions

Promotions

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.