Learn More
Thermo Scientific™ Orion™ 2 Cell Conductivity Probes
Perform applications in either the lab or the field with these versatile 2-electrode conductivity cells, available in a variety of materials and measurement ranges.
$1121.83 - $1964.15
Catalog Number | Mfr. No. | Measurement Ranges | Connector Types | For Use With (Equipment) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Measurement Ranges | Connector Types | For Use With (Equipment) | Price | Quantity | |||
13-642-223
|
Thermo Scientific™
011050MD |
1 μS/cm to 20 mS/cm | Mini-DIN | Orion Star Series Meters |
Each for $1,288.04
|
|
|||
13-642-224
|
Thermo Scientific™
011510MD |
10 μS/cm to 200 mS/cm | 8-pin Mini-DIN | Orion Star Series Meters |
Each for $1,121.83
|
|
|||
13-642-227
|
Thermo Scientific™
ORI013016MD |
0.01 to 300 μS/cm | Mini-DIN | Orion Star Series Meters |
Each for $1,964.15
|
|
|||
13-644-943
|
Thermo Scientific™
018020MD |
10 μS/cm to 2000 mS/cm | Mini-DIN | Orion Star Series Meters |
Each for $1,790.89
|
|
|||
Description
Achieve accurate measurements from ultrapure water, sea water, and more with Thermo Scientific™ Orion™ 2-Electrode Cells. These probes use two electrode cells to measure low-, standard-, and high-conductivity samples based on the cell constant. They are a versatile option for precise measurements in the lab and field. 013016MD is ideal in low ionic strength solutions, such as deionized water and ultrapure water when paired with a flow-cell assembly. 018020MD is good for chemically reactive conductivity samples, including concentrated acids and bases. 011050MD and 011510MD epoxy/graphite cells are durable, chemical resistant, and measure a wide range of conductivities in the lab or field.
The ThermoFisher Orion Conductivity electrode 013016MD complies with SM 2510B; D1125-95, -99 (A) for Drinking water measurements and USP<645> Measuring the Conductivity of Pure WaterFeatures
These probes use two electrode cells to measure low-, standard-, and high-conductivity samples based on the cell constant. They are a versatile option for precise measurements in the lab and field.
013016MD is ideal in low ionic strength solutions, such as deionized water and ultrapure water when paired with a flow-cell assembly . 018020MD is good for chemically reactive conductivity samples, including concentrated acids and bases. 011050MD and 011510MD epoxy/graphite cells are durable, chemical resistant, and measure a wide range of conductivities in the lab or field.
Solutions required
Standards
011008 100 µS/cm conductivity/TDS standard, 5 x 60 mL bottles
01100910 147 µS/cm conductivity standard, 10 pouches
011007 1413 µS/cm conductivity/TDS standard, 5 x 60 mL bottles
01100710 1413 µS/cm conductivity/TDS standard, 10 pouches
011006 12.9 mS/cm conductivity/TDS standard, 5 x 60 mL bottles
01100610 12.9 mS/cm conductivity/TDS standard, 10 pouches
011005 111 mS/cm conductivity standard, 5 x 60 mL bottles
01100510 111 mS/cm conductivity standard, 10 pouches
Conditioning solution
011001 Conditioning solution for 011050MD conductivity cells
Accessories
013017 Replacement flow cell
Items included
011050MD 011007 1413 µS/cm conductivity/TDS standard, 60 mL 1 bottle
011001 Conditioning solution, 60 mL 1 bottle
011510MD 011006 12.9 mS/cm conductivity/TDS standard, 60 mL 1 bottle
011007 1413 µS/cm conductivity/TDS standard, 60 mL 1 bottle
013016MD 011008 100 µS/cm conductivity/TDS standard, 60 mL 1 bottle
013017 Replacement flow cell
Warranty period
12 months from date of purchase
Specifications
2-Electrode Conductivity Cells |