missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
ORP Buffer 4 (246 mV), Reagents
ORP Buffer 4 (246 mV), Reagent, use for Redox. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CO0051001A
Specifications
ORP Buffer 4 (246 mV) | |
Poly Bottle | |
Colorless | |
4 | |
1.004 | |
Redox | |
Liquid |
7732-18-5,877-24-7,106-34-3 | |
1 L | |
Redox Meter | |
100°C | |
Oxidation Reduction Potential Buffer | |
Completely miscible | |
Reagent |