missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
ORP Standard, +600 mV, Ricca Chemical
$126.34 - $184.52
Chemical Identifiers
CAS | 7783-85-9 |
---|---|
Molecular Formula | FeH20N2O14S2 |
Molecular Weight (g/mol) | 392.13 |
MDL Number | MFCD00150530 |
InChI Key | MQLVWQSVRZVNIP-UHFFFAOYSA-L |
IUPAC Name | λ²-iron(2+) diammonium hexahydrate disulfate |
SMILES | [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Chemical Identifiers
7783-85-9 | |
392.13 | |
MQLVWQSVRZVNIP-UHFFFAOYSA-L | |
[NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
FeH20N2O14S2 | |
MFCD00150530 | |
λ²-iron(2+) diammonium hexahydrate disulfate |
Specifications
7783-85-9,7783-83-7,7664-93-9,7732-18-5 | |
120 mL | |
96.0,4.61,1.76,0.02 | |
FeH20N2O14S2 | |
MQLVWQSVRZVNIP-UHFFFAOYSA-L | |
[NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
392.13 | |
Oxidation-Reduction Potential Standard, +600mV vs. Ag/AgCl |
Amber Glass Bottle | |
92.0,4.43,1.69,0.02 | |
Amber | |
MFCD00150530 | |
Liquid | |
λ²-iron(2+) diammonium hexahydrate disulfate | |
4M KCl sat. with AgCl filling solution |
Safety and Handling
ShelfLife : 9 months