missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

ORP test solution, 200-275 mV, Thermo Scientific Chemicals

$108.84 - $148.84

Spécifications

Chemical Name or Material ORP test solution
Packaging Plastic-coated glass bottle
MDL Number MFCD01867801
Physical Form Liquid
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AA42131AK
Afficher les documents
Thermo Scientific Chemicals
042131AK
250 mL
chaque for $108.84
 
AA42131AP
Afficher les documents
Thermo Scientific Chemicals
042131AP
500 mL
chaque for $148.84
 
Description

Description

Test solution for ORP electrodes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

ORP test solution
Plastic-coated glass bottle
MFCD01867801
Liquid
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only