Ortho-Clinical Diagnostics™ Bovine Albumin for Blood Typing Reactions DFS Item
Enhances antibody reactions
$1269.03 - $1269.03
Specifications
Type Bovine Serum Albumin Solution
For Use With (Application) For Blood typing reactions, enhance Antigen/antibody interactions, increase in speed and sensitivity of detection
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Type Price Quantity  
Catalog Number Mfr. No. Quantity Type Price Quantity  
23287246 DFS Item
View Documents
Ortho-Clinical Diagnostics™
718280
15 x 10 mL Bovine Serum Albumin Solution
Pack of 15 Each for $1,269.03
Description
Description
  • Reagents are used to enhance antigen/antibody interactions
  • Results in an increase in speed and sensitivity of detection
Specifications
Specifications
Bovine Serum Albumin Solution
For Blood typing reactions, enhance Antigen/antibody interactions, increase in speed and sensitivity of detection
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'