Ortho-Clinical Diagnostics™ Reagent Red Cells DFS Item
Reagent red blood cells are used in the detection and identification of a variety of blood group antibodies
Supplier:  Ortho-Clinical Diagnostics™ 719201
 View more versions of this product
Catalog No. 23-287-501
Explore available promotions
Alert:
All blood products should be treated as potentially infectious. Source material from which this product was derived was found negative when tested in accordance with current FDA required tests. No known test methods can offer assurance that products derived from human blood will not transmit infectious agents.
Description
Description
  • Tests include hemagglutination and hemolysis-based results
  • A variety of testing functions are available, including:
    • Detection and/or identification of unexpected blood group antibodies
    • Affirmation of expected group antibodies
    • Serum grouping
    • Confirmation of negative antiglobulin tests
  • Screening for RhD (Rh0) positive fetal red blood cells
Specifications
Specifications
Affirmagen™
For detection and identification of a variety of Blood group Antibodies
2 x 10 mL