missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Oseltamivir phosphate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 461170010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Oseltamivir phosphate | |
White to Off-White | |
97.5% min. (HPLC) | |
Glass Bottle | |
PGZUMBJQJWIWGJ-ONAKXNSWSA-N | |
ethyl (3R,4R,5S)-4-acetamido-5-amino-3-pentan-3-yloxycyclohexene-1-carboxylate;phosphoric acid | |
410.4 | |
CHEBI:7799 | |
98% |
204255-11-8 | |
Authentic | |
C16H28N2O4·H3PO4 | |
oseltamivir phosphate, tamiflu, oseltamir phosphate, unii-4a3o49ngez, 4a3o49ngez, ethyl 3r,4r,5s-4-acetamido-5-amino-3-pentan-3-yloxy cyclohex-1-ene-1-carboxylate phosphate, ethyl 3r,4r,5s-4-acetamido-5-amino-3-1-ethylpropoxy-1-cyclohexene-1-carboxylate phosphate 1:1, osteltamivir phosphate, 3r-3alpha,4beta,5alpha-ethyl 4-acetylamino-5-amino-3-1-ethylpropoxy-1-cyclohexene-1-carboxylate phosphate 1:1 | |
CCC(CC)OC1C=C(CC(C1NC(=O)C)N)C(=O)OCC.OP(=O)(O)O | |
1 g | |
78000 | |
410.4 |
Chemical Identifiers
204255-11-8 | |
410.4 | |
oseltamivir phosphate, tamiflu, oseltamir phosphate, unii-4a3o49ngez, 4a3o49ngez, ethyl 3r,4r,5s-4-acetamido-5-amino-3-pentan-3-yloxy cyclohex-1-ene-1-carboxylate phosphate, ethyl 3r,4r,5s-4-acetamido-5-amino-3-1-ethylpropoxy-1-cyclohexene-1-carboxylate phosphate 1:1, osteltamivir phosphate, 3r-3alpha,4beta,5alpha-ethyl 4-acetylamino-5-amino-3-1-ethylpropoxy-1-cyclohexene-1-carboxylate phosphate 1:1 | |
CHEBI:7799 | |
CCC(CC)OC1C=C(CC(C1NC(=O)C)N)C(=O)OCC.OP(=O)(O)O |
C16H28N2O4·H3PO4 | |
PGZUMBJQJWIWGJ-ONAKXNSWSA-N | |
78000 | |
ethyl (3R,4R,5S)-4-acetamido-5-amino-3-pentan-3-yloxycyclohexene-1-carboxylate;phosphoric acid |
Safety and Handling
Causes serious eye irritation, Harmful if swallowed, May cause respiratory irritation, Causes skin irritation
Serious eye damage/eye irritation (category 2), Acute toxicity (category 4), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
RUO – Research Use Only