missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ospemifene 98.0+%, TCI America™
$106.65 - $252.79
Chemical Identifiers
CAS | 128607-22-7 |
---|---|
Molecular Formula | C24H23ClO2 |
Molecular Weight (g/mol) | 378.896 |
MDL Number | MFCD00871890 |
InChI Key | LUMKNAVTFCDUIE-VHXPQNKSSA-N |
PubChem CID | 3036505 |
ChEBI | CHEBI:73275 |
IUPAC Name | 2-[4-[(Z)-4-chloro-1,2-diphenylbut-1-enyl]phenoxy]ethanol |
SMILES | C1=CC=C(C=C1)C(=C(C2=CC=CC=C2)C3=CC=C(C=C3)OCCO)CCCl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
O0441100MG
|
TCI America
O0441100MG |
100 mg |
N/A
|
|
||||||
O04411G
|
TCI America
O04411G |
1 g |
N/A
|
|
||||||
Chemical Identifiers
128607-22-7 | |
378.896 | |
LUMKNAVTFCDUIE-VHXPQNKSSA-N | |
CHEBI:73275 | |
C1=CC=C(C=C1)C(=C(C2=CC=CC=C2)C3=CC=C(C=C3)OCCO)CCCl |
C24H23ClO2 | |
MFCD00871890 | |
3036505 | |
2-[4-[(Z)-4-chloro-1,2-diphenylbut-1-enyl]phenoxy]ethanol |
Specifications
128607-22-7 | |
C24H23ClO2 | |
100 mg | |
C1=CC=C(C=C1)C(=C(C2=CC=CC=C2)C3=CC=C(C=C3)OCCO)CCCl | |
378.896 | |
CHEBI:73275 | |
≥98.0% (GC) | |
Ospemifene |
White | |
MFCD00871890 | |
LUMKNAVTFCDUIE-VHXPQNKSSA-N | |
2-[4-[(Z)-4-chloro-1,2-diphenylbut-1-enyl]phenoxy]ethanol | |
3036505 | |
378.90 | |
Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator