missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite Outdoor Flammable Cabinet
Engineered to withstand the elements and potential impacts, this cabinet is OSHA, NFPA, and FM approved, ensuring your flammable chemicals are safely and securely stored outdoors.
$3417.31 - $4875.05
Specifications
| Certifications/Compliance | OSHA, NFPA, and FM approved |
|---|---|
| Color | Yellow |
| Material | Steel (18 ga) |
| Product Type | Flammables Storage Cabinet |
| For Use With (Application) | Safe outdoor storage of flammable liquids to avoid risk of potential fire hazards and chemical spills in industries like chemical manufacturing, petrochemical and laboratories |
| Catalog Number | Mfr. No. | Dimensions (D x W x H) | Door Style | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (D x W x H) | Door Style | Price | Quantity | ||||
|
19-033-905
|
Justrite
814520 |
18 x 43 x 65 in. | Self-close, Hinged, Three-point lock, Locking, Auto-close |
Each for $4,875.05
|
|
||||
|
19-033-906
|
Justrite
814500 |
18 x 43 x 65 in. | Manual-close, Hinged, Three-point lock, Locking |
Each for $4,129.46
|
|
||||
|
19-033-907
|
Justrite
813020 |
18 x 43 x 44 in. | Self-close, Hinged, Three-point lock, Locking, Auto-close |
Each for $4,038.64
|
|
||||
|
19-033-908
|
Justrite
813000 |
18 x 43 x 44 in. | Manual-close, Hinged, Three-point lock, Locking |
Each for $3,417.31
|
|
||||
Description
- Spill Slope shelves with fully welded sump
- Ground Screw
- Durable and Chemical - resistant powder coated paint
- UV resistant
- Vent with built in flash arrestors
- Welded Shelf Hangers
- Leak Proof Sump
Specifications
| OSHA, NFPA, and FM approved | |
| Steel (18 ga) | |
| Safe outdoor storage of flammable liquids to avoid risk of potential fire hazards and chemical spills in industries like chemical manufacturing, petrochemical and laboratories | |
| Adjustable |
| Yellow | |
| Flammables Storage Cabinet | |
| 2 |