VistaLab™ Technologies Ovation™ Electronic Single Channel Pipette PROMO
Ideal ergonomic choice for those needing a comprehensive menu of automated liquid handling functions.
$1469.22 - $1469.22
Specifications
Battery Type Lithium-ion
Compatible Tips VistaLab™ tip
For Use With VistaLab™ Tip
Model Right Handed
Number of Channels 1
View More Specs
Products 1
Catalog Number Mfr. No. Accuracy Increments Price Quantity  
Catalog Number Mfr. No. Accuracy Increments Price Quantity  
22-146-101TI
View Documents
VistaLab™ Technologies
10650125TI
±0.8% 0.5 μL
Each for $1,469.22
Description
Description
  • One-way disposable nozzle
  • Proprietary energy-release system discards tips at the push of a button
  • Contoured plunger electronically activates pipetting activity
  • Adjustable hook can be easily positioned for a custom fit and reduced holding effort
  • Bionatural shape gives user maximum stability when pipetting, with minimal contact pressure
  • Easy push-button controls
  • Fully electronic operations maximize efficiency for all tasks, including multiple dispensing, pipetting, reverse pipetting, mixing, serial diluting, and automatic sequential dispensing
  • Stands securely on countertops
These electronic single channel Ovation models require a power supply unit 22-147-115 (sold separately) to recharge the pipettes battery.
Electronic single channel models require an Ovation Power Supply unit (Mfr. No. 9060-9001; sold separately) to recharge the pipetter's battery
Specifications
Specifications
Lithium-ion
VistaLab™ Tip
1
Pipette
Ergonomic
5
VistaLab™ tip
Right Handed
Ovation™
LCD
VistaLab™ Tip
1 Year
SDS
missing translation for 'documents'
missing translation for 'documents'