Caframo™ Overhead Stirrer Clamp
Overhead Stirrer Clamp designed to accommodate stirrer support rods up to 16 mm (5/8 in.) diameter. Attaches overhead stirrer to Caframo and Fisherbrand stands (14-500-354 and 14-500-355).
Supplier:  Caframo™ CAFA122
Catalog No. 14501090
Explore available promotions
Description
Description
  • This clamp is an alternative to the A120 and A120FSR (14-500-6, 14-500-357)
  • Compatible with larger diameter support rods, up to 16 mm (5/8 in.)
Specifications
Specifications
Caframo™ and Fisherbrand™ Stands (14-500-354 and 14-500-355)
81 mm
40 mm
Key Only
Clamp
Cast Zinc, Aluminum