Owen Mumford™ Unistik™ 3 Single-Use Safety Lancets
Unique design features a side release button and pre-set lancet speed to ensure maximum comfort for patient and healthcare worker
Supplier:  Owen Mumford™ AT1012
 View more versions of this product
Catalog No. 22-023-381
Explore available promotions
Description
Description
  • Requires no cocking procedure – just twist off protective sterile cap and use
  • Convex platform requires less technique and training for use
  • Ergonomically designed with special indentions on each side for finger placement
  • Needle point is hidden before use and automatically retracts after use to avoid accidental needlestick injuries and cross-infection
  • Rendered inoperable after use, providing highest level of safety for healthcare workers and patients and meeting OSHA standards
  • Visual lockout feature clearly indicates that device has been used and is ready for disposal in a sharps container
  • High-flow volume suitable for cholesterol, hemoglobin and cardiac enzyme testing
US Patents 5,487,748; 6,258,112; 6,764,496
Specifications
Specifications
Single-use
21g
For ward blood glucose monitoring, HbA1c
2mm
0.07 in.
Unistik™ 3 Safety Lancet
Orange
100 Pack