Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems
Quickly and easily cast and run one or two gels in the same device with these Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems.
$2303.45 - $2901.29
Specifications
Product Line Owl
View More Specs
Products 3
Catalog Number Mfr. No. Description Volume (Metric) Buffer Price Quantity  
Catalog Number Mfr. No. Description Volume (Metric) Buffer Price Quantity  
09-518-157B
View Documents
Thermo Scientific™
P8DS1
P8DS Complete System plus Caster 300 mL
Each for $2,901.29
OW-P8DS-2
View Documents
Thermo Scientific™
P8DS2
P8DS Complete System 300 mL
Each for $2,743.05
OW-P8DS-3
View Documents
Thermo Scientific™
P8DS3
P8DS Apparatus Only 300 mL
Each for $2,303.45
Description
Description
Quickly and easily cast and run one or two gels in the same device using Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems—comprised of the P8DS, P9DS and P10DS Dual Gel Systems.
Simple, Six-Step Use:
  1. Secure the glass cassettes
  2. Pour the agarose
  3. Plug in the specially designed trough
  4. Cast polyacrylamide solution in the cassettes
  5. Load samples
  6. Run the gel
Features:
  • Manufactured using a time-tested acrylic fabrication technology, providing a durable, long-lasting system
  • Central cooling core provides even cooling across the gel surface for outstanding resolution and clear banding
  • Cooling is achieved by circulating fluid through the core with external circulator or cool tap water
  • Very simple clamping mechanism makes loading glass cassettes and most manufacturers' precast gels trouble free
Recommended for:
  • SDS electrophoresis
  • The second dimension of two-dimensional (2-D) electrophoresis
  • Agarose electrophoresis
  • Native electrophoresis
Specifications
Specifications
Owl
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'