Learn More
Oxalacetic acid, 98+%, Thermo Scientific Chemicals
$79.44 - $248.36
Chemical Identifiers
CAS | 328-42-7 |
---|---|
Molecular Formula | C4H4O5 |
Molecular Weight (g/mol) | 132.071 |
MDL Number | MFCD00002592 |
InChI Key | KHPXUQMNIQBQEV-UHFFFAOYSA-N |
Synonym | oxalacetic acid, oxaloacetic acid, 2-oxosuccinic acid, oxaloacetate, ketosuccinic acid, oxosuccinic acid, 2-ketosuccinic acid, butanedioic acid, oxo, oxobutanedioic acid, oxalacetate |
PubChem CID | 970 |
ChEBI | CHEBI:30744 |
IUPAC Name | 2-oxobutanedioic acid |
SMILES | C(C(=O)C(=O)O)C(=O)O |
Description
Oxaloacetic acid is a substrate for malate dehydrogenase and oxaloacetate decarboxylase. It is an inhibitor of succinic dehydrogenase. It is an intermediate in the citric acid cycle and glucogenesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
328-42-7 | |
132.071 | |
KHPXUQMNIQBQEV-UHFFFAOYSA-N | |
970 | |
2-oxobutanedioic acid |
C4H4O5 | |
MFCD00002592 | |
oxalacetic acid, oxaloacetic acid, 2-oxosuccinic acid, oxaloacetate, ketosuccinic acid, oxosuccinic acid, 2-ketosuccinic acid, butanedioic acid, oxo, oxobutanedioic acid, oxalacetate | |
CHEBI:30744 | |
C(C(=O)C(=O)O)C(=O)O |
Specifications
328-42-7 | |
88°C (190°F) | |
C4H4O5 | |
2 g | |
1705475 | |
oxalacetic acid, oxaloacetic acid, 2-oxosuccinic acid, oxaloacetate, ketosuccinic acid, oxosuccinic acid, 2-ketosuccinic acid, butanedioic acid, oxo, oxobutanedioic acid, oxalacetate | |
KHPXUQMNIQBQEV-UHFFFAOYSA-N | |
2-oxobutanedioic acid | |
970 | |
132.07 | |
Powder |
161°C (decomposition) | |
>98% | |
MFCD00002592 | |
UN3261 | |
14,6909 | |
Soluble in water (0.1g/ml),ether,ethanol,acetone,and acetone ethyl acetate. | |
C(C(=O)C(=O)O)C(=O)O | |
132.071 | |
CHEBI:30744 | |
≥98% | |
Oxalacetic acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 206-329-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only