missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Oxalic Acid Dihydrate (Crystalline/Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific FLA219500
Specifications
Oxalic Acid Dihydrate | |
98°C | |
White | |
C2H6O6 | |
MFCD00149102 | |
oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm | |
C(=O)(C(=O)O)O.O.O | |
126.064 | |
126.07 | |
Certified ACS | |
500 g | |
Poly Bottle | |
21.5mbar at 50°C |
1.3 | |
6153-56-6 | |
99.5 to 102.5% | |
HO2CCO2H·2H2O | |
2923 | |
GEVPUGOOGXGPIO-UHFFFAOYSA-N | |
oxalic acid;dihydrate | |
61373 | |
99.5 to 102.5% | |
4.62 | |
0.01% max. | |
Powder/Solid |
Chemical Identifiers
6153-56-6 | |
126.064 | |
GEVPUGOOGXGPIO-UHFFFAOYSA-N | |
61373 | |
C(=O)(C(=O)O)O.O.O |
C2H6O6 | |
MFCD00149102 | |
oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm | |
oxalic acid;dihydrate |
Safety and Handling
DANGER!
Emergency Overview
Causes severe eye burns. Harmful in contact with skin and if swallowed. Irritating to respiratory system. Corrosive to metals. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.
NFPA
Health:3
Flammability:1
Instability:0
DOTInformation : DOT Class 8, : Corrosive