missing translation for 'onlineSavingsMsg'
Learn More

Oxalyl chloride, 2.0M solution in dichloromethane, AcroSeal™

CAS: 79-37-8 | C2Cl2O2 | 126.93 g/mol

$114.97 - $412.23

Specifications

Chemical Name or Material Oxalyl chloride
Name Note 2.0M solution in dichloromethane
CAS 79-37-8,75-09-2
Color Colorless to Yellow
Density 1.3350g/mL
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC369181000
View Documents
Thermo Scientific Chemicals
369181000
100 mL AcroSeal™ Glass bottle
Each for $114.97
Only null left
 
AC369188000
View Documents
Thermo Scientific Chemicals
369188000
800 mL AcroSeal™ Glass Bottle
Each for $412.23
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent
Specifications

Specifications

Oxalyl chloride
79-37-8,75-09-2
1.3350g/mL
ClCOCOCl
02, 542
15, 7013
oxalyl chloride, ethanedioyl dichloride, oxalic dichloride, oxaloyl chloride, oxalic acid dichloride, oxalic acid chloride, ethanedioyl chloride, oxalylchloride, unii-r4y96317dw, cocl 2
CTSLXHKWHWQRSH-UHFFFAOYSA-N
oxalyl dichloride
65578
Liquid
2.0M solution in dichloromethane
Colorless to Yellow
C2Cl2O2
MFCD00000704
01,767; 02,301; 03,216; 04,361; 05,481; 06,424; 07,257; 08,366; 11,379; 14,150; 16,149; 17,241
1.335
Solubility in water: reacts
C(=O)(C(=O)Cl)Cl
126.93
126.93
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only