missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oxalyl Chloride 98.0+%, TCI America™

Supplier:  TCI America O0082500G

 View more versions of this product

Catalog No. O0082500G


Explore available promotions

Description

Description

Specifications

Specifications

Oxalyl Chloride
-12°C
64°C
MFCD00000704
2922
CTSLXHKWHWQRSH-UHFFFAOYSA-N
oxalyl dichloride
65578
≥98.0% (GC,T)
79-37-8
Yellow
C2Cl2O2
500 g
oxalyl chloride, ethanedioyl dichloride, oxalic dichloride, oxaloyl chloride, oxalic acid dichloride, oxalic acid chloride, ethanedioyl chloride, oxalylchloride, unii-r4y96317dw, cocl 2
C(=O)(C(=O)Cl)Cl
126.92
126.92
Liquid

Chemical Identifiers

79-37-8
126.92
CTSLXHKWHWQRSH-UHFFFAOYSA-N
65578
C(=O)(C(=O)Cl)Cl
C2Cl2O2
MFCD00000704
oxalyl chloride, ethanedioyl dichloride, oxalic dichloride, oxaloyl chloride, oxalic acid dichloride, oxalic acid chloride, ethanedioyl chloride, oxalylchloride, unii-r4y96317dw, cocl 2
oxalyl dichloride
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-2022

RTECSNumber : KI2950000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions