Learn More
Oxamic acid, 98%, Thermo Scientific Chemicals
$67.19 - $646.47
Chemical Identifiers
CAS | 471-47-6 |
---|---|
Molecular Formula | C2H3NO3 |
Molecular Weight (g/mol) | 89.05 |
MDL Number | MFCD00008006 |
InChI Key | SOWBFZRMHSNYGE-UHFFFAOYSA-N |
Synonym | 2-amino-2-oxoacetic acid, oxamate, acetic acid, aminooxo, oxalamic acid, oxamidic acid, glycine, 2-oxo, oxalic acid monoamide, amino oxo acetic acid, aminooxoacetic acid, oxamate repellent |
PubChem CID | 974 |
ChEBI | CHEBI:18058 |
IUPAC Name | oxamic acid |
SMILES | C(=O)(C(=O)O)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2034406
|
Thermo Scientific Chemicals
B2034406 |
5 g |
Each for $67.19
|
|
|||||
AAB2034414
|
Thermo Scientific Chemicals
B2034414 |
25 g |
Each for $206.36
|
|
|||||
AAB2034422
|
Thermo Scientific Chemicals
B2034422 |
100 g |
Each for $646.47
|
|
|||||
Description
Oxamic acid could also inhibit lactic dehydrogenase in the intact Ehrlich ascites tumor cell.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsOxamic acid could also inhibit lactic dehydrogenase in the intact Ehrlich ascites tumor cell.
Solubility
Soluble in water 108 mg/mL.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
471-47-6 | |
89.05 | |
SOWBFZRMHSNYGE-UHFFFAOYSA-N | |
974 | |
oxamic acid |
C2H3NO3 | |
MFCD00008006 | |
2-amino-2-oxoacetic acid, oxamate, acetic acid, aminooxo, oxalamic acid, oxamidic acid, glycine, 2-oxo, oxalic acid monoamide, amino oxo acetic acid, aminooxoacetic acid, oxamate repellent | |
CHEBI:18058 | |
C(=O)(C(=O)O)N |
Specifications
471-47-6 | |
C2H3NO3 | |
5 g | |
14,6917 | |
Soluble in water 108mg/mL. | |
C(=O)(C(=O)O)N | |
89.05 | |
CHEBI:18058 | |
98% |
∼205°C (decomposition) | |
MFCD00008006 | |
1743294 | |
2-amino-2-oxoacetic acid, oxamate, acetic acid, aminooxo, oxalamic acid, oxamidic acid, glycine, 2-oxo, oxalic acid monoamide, amino oxo acetic acid, aminooxoacetic acid, oxamate repellent | |
SOWBFZRMHSNYGE-UHFFFAOYSA-N | |
oxamic acid | |
974 | |
89.05 | |
Oxamic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 207-443-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only