missing translation for 'onlineSavingsMsg'
Learn More

Oxamic hydrazide, 99%, Thermo Scientific™

Chemical Identifiers

CAS 515-96-8
Molecular Formula C2H5N3O2
Molecular Weight (g/mol) 103.08
MDL Number MFCD00008004
InChI Key MOKRDWKSHLLYKM-UHFFFAOYSA-N
Synonym oxamic hydrazide, semioxamazide, semioxamazid, aminooxamide, oxamic acid hydrazide, oxamichydrazide, n-aminooxamide, 1-hydrazinecarbonyl formamide, oxamic acid, hydrazide, unii-bet2ubd6nv
PubChem CID 68200
IUPAC Name 2-hydrazinyl-2-oxoacetamide
SMILES C(=O)(C(=O)NN)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC129640250
SDS View Documents
Thermo Scientific Chemicals
129640250
25g Plastic bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC129641000
SDS View Documents
Thermo Scientific Chemicals
129641000
100g Plastic bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

515-96-8
103.08
MOKRDWKSHLLYKM-UHFFFAOYSA-N
68200
C(=O)(C(=O)NN)N
C2H5N3O2
MFCD00008004
oxamic hydrazide, semioxamazide, semioxamazid, aminooxamide, oxamic acid hydrazide, oxamichydrazide, n-aminooxamide, 1-hydrazinecarbonyl formamide, oxamic acid, hydrazide, unii-bet2ubd6nv
2-hydrazinyl-2-oxoacetamide

Specifications

515-96-8
99%
C2H5N3O2
MFCD00008004
02, 559
oxamic hydrazide, semioxamazide, semioxamazid, aminooxamide, oxamic acid hydrazide, oxamichydrazide, n-aminooxamide, 1-hydrazinecarbonyl formamide, oxamic acid, hydrazide, unii-bet2ubd6nv
C(=O)(C(=O)NN)N
103.08
103.08
Oxamic hydrazide, 99%
Authentic
Plastic bottle
H2NCOCONHNH2
25g
15, 8584
MOKRDWKSHLLYKM-UHFFFAOYSA-N
2-hydrazinyl-2-oxoacetamide
68200
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-213-2

RTECSNumber : AF2620000

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only