missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oxamide, 98%, Thermo Scientific Chemicals

$76.09 - $76.09

Chemical Identifiers

CAS 471-46-5
Molecular Formula C2H4N2O2
Molecular Weight (g/mol) 88.06
MDL Number MFCD00008007
InChI Key YIKSCQDJHCMVMK-UHFFFAOYSA-N
Synonym ethanediamide, oxalamide, oxamimidic acid, oxalic acid diamide, diaminoglyoxal, oxamid, amid kyseliny stavelove, formimidic acid, 1-carbamoyl, oxamid czech, unii-sbe4m0223e
PubChem CID 10113
ChEBI CHEBI:48248
IUPAC Name oxamide
SMILES C(=O)(C(=O)N)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC129651000
SDS View Documents
Thermo Scientific Chemicals
129651000
100 g Plastic bottle
Each for $76.09
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

471-46-5
88.06
YIKSCQDJHCMVMK-UHFFFAOYSA-N
10113
oxamide
C2H4N2O2
MFCD00008007
ethanediamide, oxalamide, oxamimidic acid, oxalic acid diamide, diaminoglyoxal, oxamid, amid kyseliny stavelove, formimidic acid, 1-carbamoyl, oxamid czech, unii-sbe4m0223e
CHEBI:48248
C(=O)(C(=O)N)N

Specifications

471-46-5
White
98%
C2H4N2O2
MFCD00008007
02, 545
ethanediamide, oxalamide, oxamimidic acid, oxalic acid diamide, diaminoglyoxal, oxamid, amid kyseliny stavelove, formimidic acid, 1-carbamoyl, oxamid czech, unii-sbe4m0223e
YIKSCQDJHCMVMK-UHFFFAOYSA-N
oxamide
10113
88.06
Fine Crystalline Powder
491°C
Authentic
Plastic bottle
H2NCOCONH2
100 g
15, 7017
Solubility in water: insoluble in water. Other solubilities: sparingly soluble in hot water,alcohol and ether
C(=O)(C(=O)N)N
88.06
CHEBI:48248
98%
Oxamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-442-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only