missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oxazole, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1834106

 View more versions of this product

Catalog No. AAL1834106


Explore available promotions

Description

Description

Oxazole and its derivatives are actively involved as building blocks for bio chemicals and pharmaceuticals like flopristin and darglitazone. It is also involved in industrial applications such as in dyes, fluorescent brightening agents, textile auxiliaries and plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxazole and its derivatives are actively involved as building blocks for bio chemicals and pharmaceuticals like flopristin and darglitazone. It is also involved in industrial applications such as in dyes, fluorescent brightening agents, textile auxiliaries and plastics.

Solubility
Miscible with alcohol and ether. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Oxazole
-86°C
69°C to 70°C
C3H3NO
MFCD00009751
UN2924
oxazole, unii-fjz20i1lps, fjz20i1lps, oxazol, 1,3-oxazol, pubchem8626, oxazole 1g
ZCQWOFVYLHDMMC-UHFFFAOYSA-N
1,3-oxazole
9255
69.07
288-42-6
1.05
19°C (66°F)
1.427
5 g
103851
Miscible with alcohol and ether. Slightly miscible with water.
C1=COC=N1
69.063
CHEBI:35597
≥98%

Chemical Identifiers

288-42-6
69.063
ZCQWOFVYLHDMMC-UHFFFAOYSA-N
9255
1,3-oxazole
C3H3NO
MFCD00009751
oxazole, unii-fjz20i1lps, fjz20i1lps, oxazol, 1,3-oxazol, pubchem8626, oxazole 1g
CHEBI:35597
C1=COC=N1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H225-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

EINECSNumber : 206-020-8

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only