missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Oxfendazole, Thermo Scientific™
$39.03 - $131.10
Chemical Identifiers
CAS | 53716-50-0 |
---|---|
Molecular Formula | C15H13N3O3S |
Molecular Weight (g/mol) | 315.35 |
MDL Number | 00801063 |
InChI Key | BEZZFPOZAYTVHN-UHFFFAOYNA-N |
IUPAC Name | methyl N-[6-(benzenesulfinyl)-1H-1,3-benzodiazol-2-yl]carbamate |
SMILES | COC(=O)NC1=NC2=CC=C(C=C2N1)S(=O)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC469210010
|
Thermo Scientific Chemicals
469210010 |
1 g |
Each for $39.03
|
|
|||||
AC469210050
|
Thermo Scientific Chemicals
469210050 |
5 g |
Each for $131.10
|
|
|||||
Chemical Identifiers
53716-50-0 | |
315.35 | |
BEZZFPOZAYTVHN-UHFFFAOYNA-N | |
COC(=O)NC1=NC2=CC=C(C=C2N1)S(=O)C1=CC=CC=C1 |
C15H13N3O3S | |
00801063 | |
methyl N-[6-(benzenesulfinyl)-1H-1,3-benzodiazol-2-yl]carbamate |
Specifications
53716-50-0 | |
1 g | |
Available | |
00801063 | |
BEZZFPOZAYTVHN-UHFFFAOYNA-N | |
methyl N-[6-(benzenesulfinyl)-1H-1,3-benzodiazol-2-yl]carbamate | |
Powder/Crystals |
Glass Bottle | |
White to Light Yellow to Beige | |
C15H13N3O3S | |
13-7004 | |
COC(=O)NC1=NC2=CC=C(C=C2N1)S(=O)C1=CC=CC=C1 | |
315.35 | |
Oxfendazole |
Safety and Handling
Exclamation mark
EINECSNumber : 258-714-5
RTECSNumber : FD0808000
Recommended Storage : Normal conditions