Thermo Scientific™ Oxoid™ AnaeroGen™ 2.5L Sachet
Thermo Scientific™ Oxoid AnaeroGen 2.5L Sachets are anaerobic gas generating sachets for use with Thermo Scientific™ Oxoid 2.5L jar, Part No. AG0025A (not included).
Supplier:  Thermo Scientific™ AN0025A
Catalog No. OXAN0025A
Explore available promotions
Description
Description
Thermo Scientific™ Oxoid AnaeroGen 2.5L Sachets are anaerobic gas generating sachets for use with Thermo Scientific™ Oxoid 2.5L jar, Part No. AG0025A.
  • No water or catalyst required
  • No hydrogen gas produced
Specifications
Specifications
Sachet
Thermo Scientific Oxoid
10/Pk.
2.5 L