Thermo Scientific™ Oxoid™ AnaeroGen™ 3.5L Sachet
Thermo Scientific™ Oxoid AnaeroGen 3.5L Sachet is for use with Thermo Scientific™ Oxoid Anaerobic 3.5L jars, Part No. HP0011A, HP0031A.
Supplier:  Thermo Scientific™ AN0035A
Catalog No. OXAN0035A
Explore available promotions
Description
Description
Thermo Scientific™ Oxoid AnaeroGen 3.5L Sachet is for use with Thermo Scientific™ Oxoid Anaerobic 3.5L jars, Part No. HP0011A, HP0031A.
  • No water or catalyst required
  • No hydrogen gas produced
Specifications
Specifications
Sachet
Thermo Scientific Oxoid
3.5 L
Gas Generation Sachet
10/Pk.