Thermo Scientific™ Oxoid™ AnaeroGen™ Compact
Oxoid AnaeroGen Compact is a simple anaerobic gas generator system for the anaerobic incubation of up to four petri dishes or an identification panel.
Supplier: Thermo Scientific™ AN0010C
Description
Oxoid AnaeroGen Compact is a simple anaerobic gas generator system for the anaerobic incubation of up to four petri dishes or an identification panel.
- Use 1 sachet / Compact Plastic Pouch
- Use with AnaeroGen™ Compact Sealing Clip, Part No. AN0005C
Includes: 10 sachets and 10 pouches.
Specifications
AnaeroGen Compact | |
10 sachets and 10 pouches | |
Thermo Scientific Oxoid | |
10/Pk. |