Thermo Scientific™ Oxoid™ AnaeroJar™ 2.5L
Oxoid AnaeroJar 2.5L Jar comes complete with 2.5L capacity polycarbonate jar base and lid secured to base by four self-venting clips.
Supplier:  Thermo Scientific™ AG0025A
Catalog No. OXAG0025A
Explore available promotions
Description
Description
Oxoid AnaeroJar 2.5L Jar comes complete with 2.5L capacity polycarbonate jar base and lid secured to base by four self-venting clips.
  • Lid incorporates carrying handle and vacuum release feature
  • Plate carrier included
  • Jar holds up to 12 plates
Includes:Plate carrier is included.
Specifications
Specifications
AnaeroJar
Plate carrier is included.
Each
Complete Jar
Thermo Scientific Oxoid
2.5 L
Videos
SDS
Documents
Documents
Accessories