Thermo Scientific™ Oxoid™ CO2 Gen™ Sachet
Generate carbon dioxide atmosphere in AnaeroJar™ 2.5L with Oxoid CO2 Gen Sachet.
Supplier: Thermo Scientific™ CD0025A
Description
Generate carbon dioxide atmosphere in AnaeroJar™ 2.5L with Oxoid CO2 Gen Sachet.
- No water or catalyst required
- No hydrogen gas produced
Specifications
CO2 Generating Sachet | |
Gas Generation Sachet | |
Thermo Scientific Oxoid | |
10/Pk. |