Thermo Scientific™ Oxoid™ Erythromycin Antimicrobial Susceptibility discs
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing discs.
$90.33 - $90.33
Specifications
Antimicrobial Agents Erythromycin
Description Oxoid Erythromycin Antimicrobial Susceptibility Discs
Antibiotic Code E
Product Type Antimicrobial Susceptibility Discs
Test Type Antimicrobial Susceptibility
View More Specs
Products 1
Catalog Number Mfr. No. Description Antibiotic Code Concentration Price Quantity  
Catalog Number Mfr. No. Description Antibiotic Code Concentration Price Quantity  
OXCT0020B
Accessories View Documents
Thermo Scientific™
CT0020B
Oxoid Erythromycin Antimicrobial Susceptibility Discs E 15μg
Pack of 250 Each for $90.33
Description
Description
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Erythromycin Antimicrobial Susceptibility discs.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
Erythromycin
E
Antimicrobial Susceptibility
50 Discs/Cartridge
Oxoid Erythromycin Antimicrobial Susceptibility Discs
Antimicrobial Susceptibility Discs
5 Cartridges/Pk.
50 Discs/Cartridge
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'