Thermo Scientific™ Raka-Ray Agar Base (Dehydrated)
Isolate lactic acid bacteria in beer and brewing processes with medium as recommended by ASBC and EBC.
Supplier:  Thermo Scientific™ CM0777B
Catalog No. OXCM0777B
Explore available promotions
Description
Description
Isolate lactic acid bacteria in beer and brewing processes with Thermo Scientific™ Oxoid™ Raka-Ray Agar Base (Dehydrated). The medium is based on the formula of Saha, Sondag and Middlekauff1 and is recommended by the American Society of Brewing Chemists (ASBC)2, and the European Brewing Convention (EBC)3.
Members of the family Lactobacillaceae occurring in the brewing process are important spoilage organisms because products arising from their growth and metabolism are often seriously detrimental to flavor. Detection is complicated by the diverse nutritional and environmental requirements of the family.
Raka-Ray Agar1 enables brewers to monitor in-process beer quickly and accurately for a wide range of organisms including pediococci.
Investigations in which various combinations of growth stimulating agents were added to Universal Beer Agar led to the recognition of a number of agents including sorbitan mono-oleate, liver extract, yeast extract and N-acetyl glucosamine which gave superior results in respect of colony size, colony numbers and incubation time when compared with unmodified Universal Beer Agar.
These investigations provided the basis for the formula of Raka-Ray3 Medium in which sorbitan mono-oleate is included as a stimulant for lactic acid bacteria in general4. Fructose is present as the essential carbohydrate source for Lactobacillus fructivorans5 while maltose is present to detect lactobacilli which cannot utilize glucose6.
Specifications
Specifications
Raka-Ray Agar Base
Agar
For 6.5L medium
Powder
500 g
Videos
SDS
Documents
Documents
Promotions
Promotions
For Laboratory Use Only