Thermo Scientific™ Oxoid™ X Factor Discs
Perform identification of Haemophilus spp. with Thermo Scientific™ Oxoid X Factor Discs.
Supplier:  Thermo Scientific™ DD0003T
 View more versions of this product
Catalog No. OXDD0003T
Explore available promotions
Description
Description
Perform identification of Haemophilus spp. with Thermo Scientific™ Oxoid X Factor Discs.
Ordering Alerts:
50 Discs/Cartridge
Specifications
Specifications
-10°C to -20°C
Haemophilus species
1 Cart./Pk.
X Factor Discs
X Factor Disc
50 Disks/Cartridge