Thermo Scientific™ Oxoid™ Yeast Extract Powder
Oxoid Yeast Extract Powder is a dried yeast autolysate which provides a nutritious source of nitrogen and vitamins.
$291.54 - $2203.89
Specifications
Description Yeast Extract Powder
Product Type Microbiology Media
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
OXLP0021B
View Documents
Thermo Scientific™
LP0021B
500 g
Each for $291.54
OXLP0021T
View Documents
Thermo Scientific™
LP0021T
5 kg
Each for $2,203.89
Description
Description
Oxoid Yeast Extract Powder is a dried yeast autolysate which provides a nutritious source of nitrogen and vitamins.
  • Recommended for plate count of organisms in water
  • Increases yield of organisms of many media or fermentation broths
Specifications
Specifications
Yeast Extract Powder
Microbiology Media
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'