Learn More
Oxone∣r, monopersulfate, Thermo Scientific Chemicals
$84.99 - $390.41
Chemical Identifiers
CAS | 70693-62-8 |
---|---|
Molecular Formula | H3K5O18S4 |
Molecular Weight (g/mol) | 614.74 |
MDL Number | MFCD00040551 |
InChI Key | HJKYXKSLRZKNSI-UHFFFAOYSA-I |
Synonym | Potassium peroxymonosulfate, Potassium monopersulfate triple salt |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA8989222
|
Thermo Scientific Chemicals
08989222 |
100 g |
Each for $84.99
|
|
|||||
AA89892A1
|
Thermo Scientific Chemicals
089892A1 |
1 kg |
Each for $120.23
|
|
|||||
AA89892A7
|
Thermo Scientific Chemicals
089892A7 |
5 kg |
Each for $390.41
|
|
|||||
Description
Oxone is used for halogenation of a,b-unsaturated carbonyl compounds and catalytic generation of hypervalent iodine reagents for alcohol oxidation. It is a used for rapid, and good synthesis of oxaziridines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
70693-62-8 | |
614.74 | |
HJKYXKSLRZKNSI-UHFFFAOYSA-I |
H3K5O18S4 | |
MFCD00040551 | |
Potassium peroxymonosulfate, Potassium monopersulfate triple salt |
Specifications
70693-62-8 | |
100 g | |
MFCD00040551 | |
Soluble in water (100mg/ml). | |
614.74 | |
Hygroscopic | |
Oxone |
Granules | |
H3K5O18S4 | |
Potassium peroxymonosulfate, Potassium monopersulfate triple salt | |
HJKYXKSLRZKNSI-UHFFFAOYSA-I | |
615.5 | |
1.15 g/mL |
Safety and Handling
P260-P264b-P270-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H302+H332-H314-H317-H334-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 274-778-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only