missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Aminobenzoic Acid, 99+%, MP Biomedicals

Supplier:  MP Biomedicals Inc 0219461901

 View more versions of this product

Catalog No. ICN19461901


Explore available promotions

Description

Description

Specifications

Specifications

p-Aminobenzoic Acid, 99+%
150-13-0
C7H7NO2
1 g
ALYNCZNDIQEVRV-UHFFFAOYSA-N
4-aminobenzoic acid
978
137.1
Crystalline Powder
Free Acid
White
MFCD00007894
p-aminobenzoic acid, paba, para-aminobenzoic acid, vitamin bx, sunbrella, p-carboxyaniline, 4-carboxyaniline, hachemina, paraminol, benzoic acid, 4-amino
NC1=CC=C(C=C1)C(O)=O
137.14
CHEBI:30753
Cell Culture Reagent

Chemical Identifiers

150-13-0
137.14
ALYNCZNDIQEVRV-UHFFFAOYSA-N
978
4-aminobenzoic acid
C7H7NO2
MFCD00007894
p-aminobenzoic acid, paba, para-aminobenzoic acid, vitamin bx, sunbrella, p-carboxyaniline, 4-carboxyaniline, hachemina, paraminol, benzoic acid, 4-amino
CHEBI:30753
NC1=CC=C(C=C1)C(O)=O
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room temperature

SDS
Documents

Documents

Promotions

Promotions