missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Anisaldehyde, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 104805000

 View more versions of this product

Catalog No. AC104805000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

p-Anisaldehyde
123-11-5
Colorless to Yellow
248°C to 249°C
Authentic
Glass bottle
1.5710 to 1.574
MFCD00003385
08,67
15,656
Solubility in water: 2g/L (20°C). Other solubilities: miscible withe alcohol and ether,soluble in 5 volumes of 50% alcohol,soluble in most organic solvents
COC1=CC=C(C=C1)C=O
136.15
4.1 mPa.s (25°C)
136.15
Liquid
99+%
-1°C
1.1210g/mL
116°C
99% min. (GC)
C8H8O2
CH3OC6H4CHO
500 mL
1.121
p-anisaldehyde, anisaldehyde, anisic aldehyde, p-methoxybenzaldehyde, 4-anisaldehyde, aubepine, p-anisic aldehyde, p-formylanisole, benzaldehyde, 4-methoxy, crategine
ZRSNZINYAWTAHE-UHFFFAOYSA-N
4-methoxybenzaldehyde
31244
CHEBI:28235
99+%

Chemical Identifiers

123-11-5
136.15
ZRSNZINYAWTAHE-UHFFFAOYSA-N
31244
4-methoxybenzaldehyde
C8H8O2
MFCD00003385
p-anisaldehyde, anisaldehyde, anisic aldehyde, p-methoxybenzaldehyde, 4-anisaldehyde, aubepine, p-anisic aldehyde, p-formylanisole, benzaldehyde, 4-methoxy, crategine
CHEBI:28235
COC1=CC=C(C=C1)C=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 204-602-6

RTECSNumber : BZ2625000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only