missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Benzoquinone dioxime, 95%, Thermo Scientific Chemicals

$93.67 - $1055.53

Chemical Identifiers

CAS 105-11-3
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00063636
InChI Key LNHURPJLTHSVMU-CGXWXWIYSA-N
Synonym 1,4-benzoquinone dioxime, p-benzoquinonedioxime, p-benzoquinone dioxime, p-quinone dioxime, quinone dioxime, benzoquinone dioxime, p-quinone oxime, actor q, dioxime p-benzoquinone, dibenzo pqd
PubChem CID 7737
IUPAC Name N-(4-nitrosophenyl)hydroxylamine
SMILES ON=C1C=CC(C=C1)=NO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1753218
View Documents
Thermo Scientific Chemicals
A1753218
50 g
Each for $93.67
 
AAA1753230
View Documents
Thermo Scientific Chemicals
A1753230
250 g
Each for $332.37
 
AAA175320B
View Documents
Thermo Scientific Chemicals
A175320B
1000 g
Each for $1,055.53
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

105-11-3
138.13
LNHURPJLTHSVMU-CGXWXWIYSA-N
7737
ON=C1C=CC(C=C1)=NO
C6H6N2O2
MFCD00063636
1,4-benzoquinone dioxime, p-benzoquinonedioxime, p-benzoquinone dioxime, p-quinone dioxime, quinone dioxime, benzoquinone dioxime, p-quinone oxime, actor q, dioxime p-benzoquinone, dibenzo pqd
N-(4-nitrosophenyl)hydroxylamine

Specifications

105-11-3
1.49
MFCD00063636
2043234
LNHURPJLTHSVMU-CGXWXWIYSA-N
N-(4-nitrosophenyl)hydroxylamine
7737
95%
∼223°C (decomposition)
C6H6N2O2
50 g
1,4-benzoquinone dioxime, p-benzoquinonedioxime, p-benzoquinone dioxime, p-quinone dioxime, quinone dioxime, benzoquinone dioxime, p-quinone oxime, actor q, dioxime p-benzoquinone, dibenzo pqd
ON=C1C=CC(C=C1)=NO
138.13
138.13
p-Benzoquinone dioxime
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H302
Suspected of causing cancer.
Harmful if swallowed.

P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c

H302-H351

EINECSNumber : 203-271-5

RTECSNumber : DK4900000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only