Learn More
p-Chloranil, 97%
CAS: 118-75-2 | C6Cl4O2 | 245.86 g/mol
Supplier: Thermo Scientific Chemicals A1349522
Description
p-Chloranil is used as a dye intermediate, oxidizing agent, vulcanizing agent and dehydrogenation reagent. It is also used to make chloranil electrodes for pH measurements. Further, it serves as a hydrogen acceptor and used for the aromatization reactions such as conversion of cyclohexadienes to the benzene derivatives. It is used to test the secondary amine. In addition to this, it is employed as a precursor to prepare diaziquone, which is used as a cancer chemotherapeutic agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsp-Chloranil is used as a dye intermediate, oxidizing agent, vulcanizing agent and dehydrogenation reagent. It is also used to make chloranil electrodes for pH measurements. Further, it serves as a hydrogen acceptor and used for the aromatization reactions such as conversion of cyclohexadienes to the benzene derivatives. It is used to test the secondary amine. In addition to this, it is employed as a precursor to prepare diaziquone, which is used as a cancer chemotherapeutic agent.
Solubility
Soluble in ether and acetone. Slightly soluble in chloroform, dimethylformamide, solvent naphtha, benzene, carbon disulfide and carbon tetrachloride. Insoluble in water, cold petroleum ether, methanol, dibutyl phthalate and cold alcohol.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
| p-Chloranil | |
| ∼290°C (sublimation) | |
| C6Cl4O2 | |
| 100 g | |
| 393006 | |
| chloranil, p-chloranil, tetrachloro-p-benzoquinone, tetrachloro-1,4-benzoquinone, tetrachlorobenzoquinone, spergon, coversan, reranil, vulklor, 2,3,5,6-tetrachloro-1,4-benzoquinone | |
| UGNWTBMOAKPKBL-UHFFFAOYSA-N | |
| 2,3,5,6-tetrachlorocyclohexa-2,5-diene-1,4-dione | |
| 8371 | |
| 245.88 |
| 118-75-2 | |
| 1.97 | |
| MFCD00001594 | |
| UN3077 | |
| 14,2078 | |
| Soluble in ether and acetone. Slightly soluble in chloroform,dimethylformamide,solvent naphtha,benzene,carbon disulfide and carbon tetrachloride. Insoluble in water,cold petroleum ether,methanol,dibutyl phthalate and cold alcohol. | |
| ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O | |
| 245.86 | |
| CHEBI:36703 | |
| 97% |
Chemical Identifiers
| 118-75-2 | |
| 245.86 | |
| UGNWTBMOAKPKBL-UHFFFAOYSA-N | |
| 8371 | |
| 2,3,5,6-tetrachlorocyclohexa-2,5-diene-1,4-dione |
| C6Cl4O2 | |
| MFCD00001594 | |
| chloranil, p-chloranil, tetrachloro-p-benzoquinone, tetrachloro-1,4-benzoquinone, tetrachlorobenzoquinone, spergon, coversan, reranil, vulklor, 2,3,5,6-tetrachloro-1,4-benzoquinone | |
| CHEBI:36703 | |
| ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P311-P332+P313-P362-P501c
H315-H319-H331
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 204-274-4
RTECSNumber : DK6825000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only