missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Cresol, 99+%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 405745000

 View more versions of this product

Catalog No. AC405745000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

p-Cresol
106-44-5
Colorless to Yellow
202.0°C
Authentic
Glass bottle
CH3C6H4OH
500 g
1.034
p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene
IWDCLRJOBJJRNH-UHFFFAOYSA-N
4-methylphenol
2879
CHEBI:17847
99+%
Crystalline Solid or Liquid
99+%
32.0°C to 34.0°C
1.0340g/mL
89°C
99% min. (GC)
C7H8O
MFCD00002376
06,389
15,2564
Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol,ether and chloroform
CC1=CC=C(C=C1)O
108.14
8 mPa.s (35°C)
108.14
Pure

Chemical Identifiers

106-44-5
108.14
IWDCLRJOBJJRNH-UHFFFAOYSA-N
2879
4-methylphenol
C7H8O
MFCD00002376
p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene
CHEBI:17847
CC1=CC=C(C=C1)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 203-398-6

RTECSNumber : GO6475000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only