missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Naphtholbenzein Indicator, 1% (w/v) in Titration Solvent, Ricca Chemical

$218.47 - $761.81

Specifications

Chemical Name or Material p-Naphtholbenzein Indicator
Concentration or Composition (by Analyte or Components) 1% (w/v) in Titration Solvent
Packaging Amber Glass Bottle
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
5225-4
View Documents
Ricca Chemical Company
52254
120 mL Amber Glass Bottle
Each for $218.47
 
5225-16
View Documents
Ricca Chemical Company
522516
500 mL Amber Glass Bottle
Each for $432.06
 
5225-32
View Documents
Ricca Chemical Company
522532
1 L Amber Glass Bottle
Each for $761.81
 
Description

Description

Specifications

Specifications

p-Naphtholbenzein Indicator
1% (w/v) in Titration Solvent
Amber Glass Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'