missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Nitrophenyl Phosphate,, MilliporeSigma™

Supplier:  MilliporeSigma 48766310ML

Catalog No. 48766310ML


Explore available promotions

Description

Description

Specifications

Specifications

p-Nitrophenyl Phosphate
Colorless to Yellow
10 mL
XZKIHKMTEMTJQX-UHFFFAOYSA-N
378
Liquid
330-13-2
C6H6NO6P
4-nitrophenyl phosphate, 4-nitrophenyl dihydrogen phosphate, nitrophenylphosphate, phosphoric acid, mono 4-nitrophenyl ester, p-nitrophenyl phosphate, p-nitrophenyl dihydrogen phosphate, p-nitrophenol phosphate, p-nitrophenylphosphate, pnpp
219.09
CHEBI:17440

Chemical Identifiers

330-13-2
219.09
4-nitrophenyl phosphate, 4-nitrophenyl dihydrogen phosphate, nitrophenylphosphate, phosphoric acid, mono 4-nitrophenyl ester, p-nitrophenyl phosphate, p-nitrophenyl dihydrogen phosphate, p-nitrophenol phosphate, p-nitrophenylphosphate, pnpp
CHEBI:17440
C6H6NO6P
XZKIHKMTEMTJQX-UHFFFAOYSA-N
378
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 4°C (39.2°F).

SDS
Documents

Documents

Promotions

Promotions