missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Quinquephenyl, 98%, Thermo Scientific Chemicals

$717.68 - $717.68

Chemical Identifiers

CAS 3073-05-0
Molecular Formula C30H22
Molecular Weight (g/mol) 382.506
MDL Number MFCD00059012
InChI Key OMCUOJTVNIHQTI-UHFFFAOYSA-N
Synonym p-quinquephenyl, p-pentaphenyl, 1,1':4',1:4,1':4',1-quinquephenyl, p-quinquiphenyl, acmc-209hie, 1,4-bis 4-phenylphenyl benzene, 1-phenyl-4-4-4-phenylphenyl phenyl benzene, 1,1:4,1':4',1-quinquephenyl, 4-1,1'-biphenyl-4-yl-4'-phenyl-1,1'-biphenyl
PubChem CID 137813
IUPAC Name 1,4-bis(4-phenylphenyl)benzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)C4=CC=C(C=C4)C5=CC=CC=C5
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2630803
View Documents
Thermo Scientific Chemicals
H2630803
1 g
Each for $717.68
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3073-05-0
382.506
OMCUOJTVNIHQTI-UHFFFAOYSA-N
137813
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)C4=CC=C(C=C4)C5=CC=CC=C5
C30H22
MFCD00059012
p-quinquephenyl, p-pentaphenyl, 1,1':4',1:4,1':4',1-quinquephenyl, p-quinquiphenyl, acmc-209hie, 1,4-bis 4-phenylphenyl benzene, 1-phenyl-4-4-4-phenylphenyl phenyl benzene, 1,1:4,1':4',1-quinquephenyl, 4-1,1'-biphenyl-4-yl-4'-phenyl-1,1'-biphenyl
1,4-bis(4-phenylphenyl)benzene

Specifications

3073-05-0
MFCD00059012
p-quinquephenyl, p-pentaphenyl, 1,1':4',1:4,1':4',1-quinquephenyl, p-quinquiphenyl, acmc-209hie, 1,4-bis 4-phenylphenyl benzene, 1-phenyl-4-4-4-phenylphenyl phenyl benzene, 1,1:4,1':4',1-quinquephenyl, 4-1,1'-biphenyl-4-yl-4'-phenyl-1,1'-biphenyl
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)C4=CC=C(C=C4)C5=CC=CC=C5
382.506
382.5
p-Quinquephenyl
C30H22
1 g
OMCUOJTVNIHQTI-UHFFFAOYSA-N
1,4-bis(4-phenylphenyl)benzene
137813
98%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only