missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Sexiphenyl, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2658603

 View more versions of this product

Catalog No. AAH2658603


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

p-Sexiphenyl
>400°C
MFCD00039559
p-sexiphenyl, p-hexaphenyl, p-quaterphenyl, 4,4'-diphenyl, 1-phenyl-4-4-4-4-phenylphenyl phenyl phenyl benzene, p-sexiphenylene, acmc-209k0d, phthalocyanine lead ii purified by sublimation, 1,1':4',1:4,1'-quaterphenyl, 4,4'-diphenyl, 4,4'-bis 1,1'-biphenyl-4-yl-1,1'-biphenyl
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)C4=CC=C(C=C4)C5=CC=C(C=C5)C6=CC=CC=C6
458.604
458.59
4499-83-6
C36H26
1 g
ZEMDSNVUUOCIED-UHFFFAOYSA-N
1-phenyl-4-[4-[4-(4-phenylphenyl)phenyl]phenyl]benzene
78254
95%

Chemical Identifiers

4499-83-6
458.604
ZEMDSNVUUOCIED-UHFFFAOYSA-N
78254
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)C4=CC=C(C=C4)C5=CC=C(C=C5)C6=CC=CC=C6
C36H26
MFCD00039559
p-sexiphenyl, p-hexaphenyl, p-quaterphenyl, 4,4'-diphenyl, 1-phenyl-4-4-4-4-phenylphenyl phenyl phenyl benzene, p-sexiphenylene, acmc-209k0d, phthalocyanine lead ii purified by sublimation, 1,1':4',1:4,1'-quaterphenyl, 4,4'-diphenyl, 4,4'-bis 1,1'-biphenyl-4-yl-1,1'-biphenyl
1-phenyl-4-[4-[4-(4-phenylphenyl)phenyl]phenyl]benzene
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 224-799-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only