Learn More
Thermo Scientific Chemicals p-Terphenyl, 99+%, pure
$69.32 - $69.32
Chemical Identifiers
CAS | 92-94-4 |
---|---|
Molecular Formula | C18H14 |
Molecular Weight (g/mol) | 230.31 |
MDL Number | MFCD00003061 |
InChI Key | XJKSTNDFUHDPQJ-UHFFFAOYSA-N |
Synonym | p-terphenyl, 1,1':4',1-terphenyl, p-diphenylbenzene, p-triphenyl, santowax p, 4-phenylbiphenyl, 4-phenyldiphenyl, para-terphenyl, biphenyl, 4-phenyl, pyrogallol tannin |
PubChem CID | 7115 |
ChEBI | CHEBI:52242 |
IUPAC Name | 1,4-diphenylbenzene |
SMILES | C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC137920250
|
Thermo Scientific Chemicals
137920250 |
25 g | Plastic bottle |
Each for $69.32
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
92-94-4 | |
230.31 | |
XJKSTNDFUHDPQJ-UHFFFAOYSA-N | |
7115 | |
1,4-diphenylbenzene |
C18H14 | |
MFCD00003061 | |
p-terphenyl, 1,1':4',1-terphenyl, p-diphenylbenzene, p-triphenyl, santowax p, 4-phenylbiphenyl, 4-phenyldiphenyl, para-terphenyl, biphenyl, 4-phenyl, pyrogallol tannin | |
CHEBI:52242 | |
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3 |
Specifications
92-94-4 | |
White to Beige | |
383.0°C | |
Authentic | |
Plastic bottle | |
C6H5C6H4C6H5 | |
25 g | |
p-terphenyl, 1,1':4',1-terphenyl, p-diphenylbenzene, p-triphenyl, santowax p, 4-phenylbiphenyl, 4-phenyldiphenyl, para-terphenyl, biphenyl, 4-phenyl, pyrogallol tannin | |
XJKSTNDFUHDPQJ-UHFFFAOYSA-N | |
1,4-diphenylbenzene | |
7115 | |
230.31 | |
Pure | |
p-Terphenyl, Pure |
209.0°C to 216.0°C | |
207g/mL | |
207°C | |
99% min. (GC) | |
C18H14 | |
MFCD00003061 | |
05, 695 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,benzene and toluene | |
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3 | |
230.31 | |
CHEBI:52242 | |
99+% | |
Crystalline Powder, Shiny Crystals or Flakes |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
GHS Signal Word: Warning
EINECSNumber : 202-205-2
RUO – Research Use Only