missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Terphenyl, 99+%, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals A1483314

 View more versions of this product

Catalog No. AAA1483314


Explore available promotions

Description

Description

p-Terphenyl is a specialty material, and may be used in ionized radiation detectors, nonpolar laser dye, and single molecule optical probe of scanning near-field microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Terphenyl is a specialty material, and may be used in ionized radiation detectors, nonpolar laser dye, and single molecule optical probe of scanning near-field microscopy.

Solubility
Insoluble in water. Soluble in hot benzene. Very soluble in hot ethyl alcohol. 

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Specifications

p-Terphenyl
212°C to 215°C
405°C
C18H14
25 g
p-terphenyl, 1,1':4',1-terphenyl, p-diphenylbenzene, p-triphenyl, santowax p, 4-phenylbiphenyl, 4-phenyldiphenyl, para-terphenyl, biphenyl, 4-phenyl, pyrogallol tannin
XJKSTNDFUHDPQJ-UHFFFAOYSA-N
1,4-diphenylbenzene
7115
230.31
92-94-4
1.23
207°C (404°F)
MFCD00003061
1908447
Insoluble in water. Soluble in hot benzene. Very soluble in hot ethyl alcohol.
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3
230.31
CHEBI:52242
≥99%

Chemical Identifiers

92-94-4
230.31
XJKSTNDFUHDPQJ-UHFFFAOYSA-N
7115
1,4-diphenylbenzene
C18H14
MFCD00003061
p-terphenyl, 1,1':4',1-terphenyl, p-diphenylbenzene, p-triphenyl, santowax p, 4-phenylbiphenyl, 4-phenyldiphenyl, para-terphenyl, biphenyl, 4-phenyl, pyrogallol tannin
CHEBI:52242
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 202-205-2

RTECSNumber : WZ6475000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only