Learn More
p-Terphenyl, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1483322
Description
p-Terphenyl is a specialty material, and may be used in ionized radiation detectors, nonpolar laser dye, and single molecule optical probe of scanning near-field microscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsp-Terphenyl is a specialty material, and may be used in ionized radiation detectors, nonpolar laser dye, and single molecule optical probe of scanning near-field microscopy.
Solubility
Insoluble in water. Soluble in hot benzene. Very soluble in hot ethyl alcohol.Â
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
p-Terphenyl | |
212°C to 215°C | |
405°C | |
C18H14 | |
100 g | |
p-terphenyl, 1,1':4',1-terphenyl, p-diphenylbenzene, p-triphenyl, santowax p, 4-phenylbiphenyl, 4-phenyldiphenyl, para-terphenyl, biphenyl, 4-phenyl, pyrogallol tannin | |
XJKSTNDFUHDPQJ-UHFFFAOYSA-N | |
1,4-diphenylbenzene | |
7115 | |
230.31 |
92-94-4 | |
1.23 | |
207°C (404°F) | |
MFCD00003061 | |
1908447 | |
Insoluble in water. Soluble in hot benzene. Very soluble in hot ethyl alcohol. | |
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3 | |
230.31 | |
CHEBI:52242 | |
≥99% |
Chemical Identifiers
92-94-4 | |
230.31 | |
XJKSTNDFUHDPQJ-UHFFFAOYSA-N | |
7115 | |
1,4-diphenylbenzene |
C18H14 | |
MFCD00003061 | |
p-terphenyl, 1,1':4',1-terphenyl, p-diphenylbenzene, p-triphenyl, santowax p, 4-phenylbiphenyl, 4-phenyldiphenyl, para-terphenyl, biphenyl, 4-phenyl, pyrogallol tannin | |
CHEBI:52242 | |
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 202-205-2
RTECSNumber : WZ6475000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only