Learn More
p-Tolualdehyde, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1138406
Description
p-Tolualdehyde is used as an intermediate for the synthesis of pharmaceuticals, dyes perfumes and agrochemicals. It is also used as a fixative of flavorings. It is also used as an important organic intermediates, used for spices, triphenylmethane dye synthesis, etc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsp-Tolualdehyde is used as an intermediate for the synthesis of pharmaceuticals, dyes perfumes and agrochemicals. It is also used as a fixative of flavorings. It is also used as an important organic intermediates, used for spices, triphenylmethane dye synthesis, etc.
Solubility
It is slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Incompatible to Strong bases, Strong oxidizing agents, Strong reducing agents
Specifications
p-Tolualdehyde | |
-6°C | |
204°C | |
C8H8O | |
MFCD00006954 | |
385772 | |
p-tolualdehyde, 4-tolualdehyde, p-formyltoluene, p-tolylaldehyde, p-toluylaldehyde, p-methylbenzaldehyde, para-tolualdehyde, benzaldehyde, 4-methyl, para-methylbenzaldehyde, para-toluyl aldehyde | |
FXLOVSHXALFLKQ-UHFFFAOYSA-N | |
4-methylbenzaldehyde | |
7725 | |
120.15 |
104-87-0 | |
1.018 | |
80°C (176°F) | |
1.546 | |
5 g | |
Air Sensitive | |
It is slightly soluble in water. | |
CC1=CC=C(C=C1)C=O | |
120.151 | |
CHEBI:28617 | |
98% |
Chemical Identifiers
104-87-0 | |
120.151 | |
FXLOVSHXALFLKQ-UHFFFAOYSA-N | |
7725 | |
4-methylbenzaldehyde |
C8H8O | |
MFCD00006954 | |
p-tolualdehyde, 4-tolualdehyde, p-formyltoluene, p-tolylaldehyde, p-toluylaldehyde, p-methylbenzaldehyde, para-tolualdehyde, benzaldehyde, 4-methyl, para-methylbenzaldehyde, para-toluyl aldehyde | |
CHEBI:28617 | |
CC1=CC=C(C=C1)C=O |
Safety and Handling
GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302-H315-H319-H335
EINECSNumber : 203-246-9
RTECSNumber : CU7034500
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only