Learn More
p-Tolualdehyde, 99+%, Thermo Scientific Chemicals
$75.43 - $246.46
Chemical Identifiers
CAS | 104-87-0 |
---|---|
Molecular Formula | C8H8O |
Molecular Weight (g/mol) | 120.15 |
MDL Number | MFCD00006954 |
InChI Key | FXLOVSHXALFLKQ-UHFFFAOYSA-N |
Synonym | p-tolualdehyde, 4-tolualdehyde, p-formyltoluene, p-tolylaldehyde, p-toluylaldehyde, p-methylbenzaldehyde, para-tolualdehyde, benzaldehyde, 4-methyl, para-methylbenzaldehyde, para-toluyl aldehyde |
PubChem CID | 7725 |
ChEBI | CHEBI:28617 |
IUPAC Name | 4-methylbenzaldehyde |
SMILES | CC1=CC=C(C=C1)C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC421141000
|
Thermo Scientific Chemicals
421141000 |
100 g | Glass bottle |
Each for $75.43
|
|
|||||
AC421145000
|
Thermo Scientific Chemicals
421145000 |
500 g | Glass bottle |
Each for $246.46
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
104-87-0 | |
120.15 | |
FXLOVSHXALFLKQ-UHFFFAOYSA-N | |
7725 | |
4-methylbenzaldehyde |
C8H8O | |
MFCD00006954 | |
p-tolualdehyde, 4-tolualdehyde, p-formyltoluene, p-tolylaldehyde, p-toluylaldehyde, p-methylbenzaldehyde, para-tolualdehyde, benzaldehyde, 4-methyl, para-methylbenzaldehyde, para-toluyl aldehyde | |
CHEBI:28617 | |
CC1=CC=C(C=C1)C=O |
Specifications
104-87-0 | |
Colorless to Yellow | |
204.0°C to 205.0°C | |
Authentic | |
Glass bottle | |
1.5420 to 1.5480 | |
MFCD00006954 | |
07,297 | |
p-tolualdehyde, 4-tolualdehyde, p-formyltoluene, p-tolylaldehyde, p-toluylaldehyde, p-methylbenzaldehyde, para-tolualdehyde, benzaldehyde, 4-methyl, para-methylbenzaldehyde, para-toluyl aldehyde | |
FXLOVSHXALFLKQ-UHFFFAOYSA-N | |
4-methylbenzaldehyde | |
7725 | |
120.15 | |
Liquid |
-6.0°C | |
1.0150g/mL | |
71°C | |
99% min. (GC) | |
C8H8O | |
CH3C6H4CHO | |
100 g | |
1.015 | |
Solubility in water: 0.25g/l (25°C). Other solubilities: soluble in alcohol and ether | |
CC1=CC=C(C=C1)C=O | |
120.15 | |
CHEBI:28617 | |
99+% | |
p-Tolualdehyde, 99+% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
GHS Signal Word: Warning
EINECSNumber : 203-246-9
RTECSNumber : CU7034500
TSCA : TSCA
RUO – Research Use Only