missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Toluamide, 98+%, Thermo Scientific Chemicals

$62.63 - $628.21

Chemical Identifiers

CAS 619-55-6
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00008002
InChI Key UHBGYFCCKRAEHA-UHFFFAOYSA-N
Synonym p-toluamide, benzamide, 4-methyl, p-methylbenzamide, 4-methyl-benzamide, 4-toluamide, 4-carbamoyltoluene, unii-n0v326v2f5, tolylsaureamid, n-toluamide, p-tolylamide
PubChem CID 69274
IUPAC Name 4-methylbenzamide
SMILES CC1=CC=C(C=C1)C(N)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1155914
View Documents
Thermo Scientific Chemicals
A1155914
25 g
Each for $62.63
 
AAA1155922
View Documents
Thermo Scientific Chemicals
A1155922
100 g
Each for $150.66
 
AAA1155936
View Documents
Thermo Scientific Chemicals
A1155936
500 g
Each for $628.21
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

619-55-6
135.17
UHBGYFCCKRAEHA-UHFFFAOYSA-N
69274
CC1=CC=C(C=C1)C(N)=O
C8H9NO
MFCD00008002
p-toluamide, benzamide, 4-methyl, p-methylbenzamide, 4-methyl-benzamide, 4-toluamide, 4-carbamoyltoluene, unii-n0v326v2f5, tolylsaureamid, n-toluamide, p-tolylamide
4-methylbenzamide

Specifications

619-55-6
C8H9NO
25 g
p-toluamide, benzamide, 4-methyl, p-methylbenzamide, 4-methyl-benzamide, 4-toluamide, 4-carbamoyltoluene, unii-n0v326v2f5, tolylsaureamid, n-toluamide, p-tolylamide
CC1=CC=C(C=C1)C(N)=O
135.17
135.17
p-Toluamide
158°C to 160°C
MFCD00008002
774353
UHBGYFCCKRAEHA-UHFFFAOYSA-N
4-methylbenzamide
69274
≥98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 210-600-6

RTECSNumber : CV5570150

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only