Learn More
p-Toluenesulfonamide, 99%
CAS: 70-55-3 | C7H9NO2S | 171.21 g/mol
$54.59 - $145.83
Chemical Identifiers
| CAS | 70-55-3 |
|---|---|
| Molecular Formula | C7H9NO2S |
| Molecular Weight (g/mol) | 171.21 |
| MDL Number | MFCD00011692 |
| InChI Key | LMYRWZFENFIFIT-UHFFFAOYSA-N |
| Synonym | p-toluenesulfonamide, 4-toluenesulfonamide, tosylamide, p-tosylamide, benzenesulfonamide, 4-methyl, p-tolylsulfonamide, toluene-4-sulfonamide, tolylsulfonamide, 4-methylbenzene-1-sulfonamide, p-toluenesulfamide |
| PubChem CID | 6269 |
| ChEBI | CHEBI:34435 |
| IUPAC Name | 4-methylbenzenesulfonamide |
| SMILES | CC1=CC=C(C=C1)S(=O)(=O)N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC147692500
|
Thermo Scientific Chemicals
147692500 |
250 g | Glass bottle |
Each for $54.59
|
|
||||
|
AC147690010
|
Thermo Scientific Chemicals
147690010 |
1 kg | Glass bottle |
Each for $145.83
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 70-55-3 | |
| 171.21 | |
| LMYRWZFENFIFIT-UHFFFAOYSA-N | |
| 6269 | |
| 4-methylbenzenesulfonamide |
| C7H9NO2S | |
| MFCD00011692 | |
| p-toluenesulfonamide, 4-toluenesulfonamide, tosylamide, p-tosylamide, benzenesulfonamide, 4-methyl, p-tolylsulfonamide, toluene-4-sulfonamide, tolylsulfonamide, 4-methylbenzene-1-sulfonamide, p-toluenesulfamide | |
| CHEBI:34435 | |
| CC1=CC=C(C=C1)S(=O)(=O)N |
Specifications
| 70-55-3 | |
| White | |
| 202°C | |
| 98.5% min. (HPLC) | |
| C7H9NO2S | |
| MFCD00011692 | |
| 11, 104 | |
| p-toluenesulfonamide, 4-toluenesulfonamide, tosylamide, p-tosylamide, benzenesulfonamide, 4-methyl, p-tolylsulfonamide, toluene-4-sulfonamide, tolylsulfonamide, 4-methylbenzene-1-sulfonamide, p-toluenesulfamide | |
| LMYRWZFENFIFIT-UHFFFAOYSA-N | |
| 4-methylbenzenesulfonamide | |
| 6269 | |
| 171.21 | |
| Crystalline Powder |
| 136.0°C to 140.0°C | |
| 221.0°C (10.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| CH3C6H4SO2NH2 | |
| 250 g | |
| 13,294 | |
| Solubility in water: 0.32 g/100 mL(25° c). Other solubilities: soluble in alcohol | |
| CC1=CC=C(C=C1)S(=O)(=O)N | |
| 171.21 | |
| CHEBI:34435 | |
| 99% | |
| p-Toluenesulfonamide |
Safety and Handling
GHS H Statement
Harmful in contact with skin. May cause respiratory irritation. Harmful to aquatic life with long lasting effects. Causes skin irritation. Harmful if swallowed. Causes serious eye irritation. Harmful if inhaled. Contact with acids liberates very toxic gas.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Keep container tightly closed. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 200-741-1
RUO – Research Use Only